(R)-tert-Butyl-2-amino-3-phenylpropanoate hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: D-Phenylalanine tert-butyl ester hydrochloride, H-D-Phe-OtBu·HCl, H-D-Phe-OtBu.HCl

075724-1G 075724-5G 075724-10G
102542-718EA 107.69 USD
102542-718 102542-720 102542-722
(R)-tert-Butyl-2-amino-3-phenylpropanoate hydrochloride ≥95%
(R)-tert-Butyl-2-amino-3-phenylpropanoate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉NO₂·HCl
MDL Number: MFCD00153455
CAS Number: 3403-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR