5-Amino-1H-imidazole-4-carboxamide hydrochloride ≥95%

Supplier: Matrix Scientific

075662-100G
102542-406EA 156.92 USD
102542-406
5-Amino-1H-imidazole-4-carboxamide hydrochloride ≥95%
5-Amino-1H-imidazole-4-carboxamide hydrochloride

Hazard: Irritant/Hygroscopic/Stored under Argon

Formula: C₄H₇ClN₄O
Melting Pt: 250…252 °C
MDL Number: MFCD00012704
CAS Number: 72-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR