(R)-2-Amino-3-(tritylthio)propanamide ≥95%

Supplier: Matrix Scientific

Synonyms: H-Cys(Trt)-NH2, H-Cys(Trt)-NH₂, S-Trityl-L-cysteinamide

075659-1G 075659-5G 075659-10G
102542-390EA 178.46 USD
102542-390 102542-392 102542-394
(R)-2-Amino-3-(tritylthio)propanamide ≥95%
(R)-2-Amino-3-(tritylthio)propanamide

Hazard: Irritant

Formula: C₂₂H₂₂N₂OS
MW: 362.49 g/mol
MDL Number: MFCD22126061
CAS Number: 166737-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR