Fmoc-Glu-OtBu ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-4-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(tert-butoxy)-5-oxopentanoic acid, N-Fmoc-L-glutamic acid 1-tert-butyl ester, Fmoc-Glu-OtBu

075631-5G
102542-244EA 58.46 USD
102542-244
Fmoc-Glu-OtBu ≥95%
Fmoc-Glu-OtBu

Hazard: Irritant

Formula: C₂₄H₂₇NO₆
MW: 425.47 g/mol
MDL Number: MFCD00065648
CAS Number: 84793-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR