5-Chloro-2-nitropyridine ≥95%

Supplier: Matrix Scientific

075557-1G 075557-5G 075557-10G
102541-898EA 15.38 USD
102541-898 102541-900 102541-902
5-Chloro-2-nitropyridine ≥95%
5-Chloro-2-nitropyridine

Hazard: Irritant

Formula: C₅H₃ClN₂O₂
MDL Number: MFCD03092916
CAS Number: 52092-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR