5-Chloro-2-nitropyridine ≥95%
Supplier: Matrix Scientific
075557-1G
075557-5G
075557-10G
102541-898EA
15.38
USD
102541-898
102541-900
102541-902
5-Chloro-2-nitropyridine ≥95%
5-Chloro-2-nitropyridine
Hazard: Irritant
Formula:
C₅H₃ClN₂O₂ |
MDL Number:
MFCD03092916 CAS Number: 52092-47-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...