L(+)-Arabinose ≥95%

Supplier: Matrix Scientific

Synonyms: Pectinose, Pectin Sugar

075542-100G
102541-832EA 123.08 USD
102541-832
L(+)-Arabinose ≥95%
L(+)-Arabinose

Hazard: Irritant

Formula: C₅H₁₀O₅
MW: 150.13 g/mol
MDL Number: MFCD00067709
CAS Number: 5328-37-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ad925ac21d4f12371621b1c44dbb6cda0a12da8&vid=6b9738c515009e5720388ea307490836c3b8afa8&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR