((3-Bromopropoxy)methyl)benzene 95

Supplier: Matrix Scientific

Synonyms: Benzyl 3-bromopropyl ether, 3-bromopropylbenzyl ether

075513-5G
102541-702EA 158.46 USD
102541-702
((3-Bromopropoxy)methyl)benzene 95
((3-Bromopropoxy)methyl)benzene

Hazard: Irritant

Formula: C₁₀H₁₃BrO
MW: 229.117 g/mol
Boiling Pt: 130…132 °C (8 mmHg)
Density: 1.298
Flash Pt: 113 °C
MDL Number: MFCD00134570
CAS Number: 54314-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR