N,N'-Bis-Z-1-guanylpyrazole ≥95%

Supplier: Matrix Scientific

Synonyms: N,N'-Bis-Z-1-Guanylpyrazole

075487-5G
102541-576EA 116.92 USD
102541-576
N,N'-Bis-Z-1-guanylpyrazole ≥95%
N,N'-Bis-Z-1-guanylpyrazole

Hazard: Irritant

Formula: C₂₀H₁₈N₄O₄
MW: 378.38 g/mol
MDL Number: MFCD02683516
CAS Number: 152120-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR