2-Bromo-3-chloroaniline ≥95%

Supplier: Matrix Scientific

075474-250MG 075474-1G 075474-5G
102541-520EA 128.15 USD
102541-520 102541-522 102541-524
2-Bromo-3-chloroaniline ≥95%
2-Bromo-3-chloroaniline

Hazard: Irritant

Formula: C₆H₅BrClN
MDL Number: MFCD10699543
CAS Number: 96558-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR