Sarcosine ≥95%

Supplier: Matrix Scientific

075461-100G
102541-462EA 64.62 USD
102541-462
Sarcosine ≥95%
Sarcosine

Hazard: Irritant

Formula: C₃H₇NO₂
MDL Number: MFCD00004279
CAS Number: 107-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR