(S)-2-Amino-4-phenylbutanoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: H-HoPhe-OH, H-Homophe-OH, (S)-2-Amino-4-phenylbutyric Acid

075436-100G
102541-348EA 129.23 USD
102541-348
(S)-2-Amino-4-phenylbutanoic acid ≥95%
(S)-2-Amino-4-phenylbutanoic acid

Hazard: Irritant

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
MDL Number: MFCD00002619
CAS Number: 943-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR