3-Chlorophenyl acetylene ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Chloro-3-ethynylbenzene, 3-Chlorophenylacetylene, (3-Chlorophenyl)acetylene

075416-1G 075416-5G 075416-10G
102541-254EA 15.38 USD
102541-254 102541-256 102541-258
3-Chlorophenyl acetylene ≥95%
3-Chlorophenyl acetylene

Hazard: Irritant

Formula: C₈H₅Cl
MW: 136.578 g/mol
Boiling Pt: 178…179 °C
Density: 1.109
Flash Pt: 62.78 °C
MDL Number: MFCD03839984
CAS Number: 766-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR