4-Amino-2,2,6,6-tetramethylpiperidine ≥95%

Supplier: Matrix Scientific

075353-10G 075353-100G
102540-966EA 64.62 USD
102540-966 102540-968
4-Amino-2,2,6,6-tetramethylpiperidine ≥95%
4-Amino-2,2,6,6-tetramethylpiperidine

Hazard: Irritant

Formula: C₉H₂₀N₂
MDL Number: MFCD00005984
CAS Number: 36768-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR