L-Phenylalanine ≥95%
Supplier: Matrix Scientific
075335-100G
075335-500G
102540-888EA
60
USD
102540-888
102540-890
L-Phenylalanine ≥95%
L-α-Phenylalanine
Hazard: Irritant
Formula:
C₉H₁₁NO₂ Melting Pt: 227 °C. (dec.) |
MDL Number:
MFCD00064227 CAS Number: 63-91-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...