L-Phenylalanine ≥95%

Supplier: Matrix Scientific

075335-100G 075335-500G
102540-888EA 60 USD
102540-888 102540-890
L-Phenylalanine ≥95%
L-α-Phenylalanine

Hazard: Irritant

Formula: C₉H₁₁NO₂
Melting Pt: 227 °C. (dec.)
MDL Number: MFCD00064227
CAS Number: 63-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR