L(+)-Ornithine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: H-Orn-OH Hydrochloride, H-Orn-OH·HCl

075326-100G
102540-854EA 72.31 USD
102540-854
L(+)-Ornithine hydrochloride ≥95%
L(+)-Ornithine hydrochloride

Hazard: Irritant

Formula: C₅H₁₂N₂O₂·HCl
MW: 168.62 g/mol
MDL Number: MFCD00064562
CAS Number: 3184-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR