1-Methylcyclopentanol ≥95%
Supplier: Matrix Scientific
075294-1G
075294-5G
075294-10G
102540-706EA
33.85
USD
102540-706
102540-708
102540-710
1-Methylcyclopentanol ≥95%
1-Methylcyclopentanol
Hazard: Irritant
Formula:
C₆H₁₂O |
MDL Number:
MFCD00001364 CAS Number: 1462-03-9 UN: 1325 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...