L-(+)-Methyl-tert-leucinate hydrochloride ≥95%

Supplier: Matrix Scientific

075292-1G 075292-5G
102540-696EA 26.15 USD
102540-696 102540-698
L-(+)-Methyl-tert-leucinate hydrochloride ≥95%
L(+)-Methyl tert-leucinate hydrochloride

Hazard: Irritant

Formula: C₇H₁₅NO₂·HCl
Melting Pt: 183 °C
MDL Number: MFCD00040604
CAS Number: 63038-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR