Trimethylsilanol ≥95%

Supplier: Matrix Scientific

075262-100G
102540-556EA 453.85 USD
102540-556
Trimethylsilanol ≥95%
Trimethylsilanol

TCA listed: T
Hazard: Flammable

Formula: C₃H₁₀OSi
Boiling Pt: 98…99 °C
Density: 0.8144
Flash Pt: 4 °C
MDL Number: MFCD02751657
CAS Number: 1066-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR