4-Phenylbenzophenone ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Benzoylbiphenyl

075242-10G 075242-100G
102540-460EA 64.62 USD
102540-460 102540-462
4-Phenylbenzophenone ≥95%
4-Phenylbenzophenone

Hazard: Irritant

Formula: C₁₉H₁₄O
MDL Number: MFCD00003079
CAS Number: 2128-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR