5,6-Diaminopyrimidine-2,4-diol xsulfate ≥95%

Supplier: Matrix Scientific

Synonyms: 5,6-Diaminouracil sulfate, 5,6-Diaminouracil sulphate

075167-5G
102540-102EA 304.62 USD
102540-102
5,6-Diaminopyrimidine-2,4-diol xsulfate ≥95%
5,6-Diaminopyrimidine-2,4-diol xsulfate

Hazard: Irritant

Formula: C₄H₈N₄O₆S
MW: 240.20 g/mol
MDL Number: MFCD00023155
CAS Number: 32014-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR