4-Bromomethyl-2-cyanobiphenyl ≥95%

Supplier: Matrix Scientific

Synonyms: 4'-(Bromomethyl)-[1,1'-biphenyl]-2-carbonitrile

075139-5G 075139-10G 075139-100G
102539-960EA 107.69 USD
102539-960 102539-962 102539-964
4-Bromomethyl-2-cyanobiphenyl ≥95%
4-Bromomethyl-2-cyanobiphenyl

Hazard: Irritant

Formula: C₁₄H₁₀BrN
MW: 272.14 g/mol
MDL Number: MFCD00671503
CAS Number: 114772-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR