2-Hydroxy-m-anisaldehyde ≥95%

Supplier: Matrix Scientific

075125-100G
102539-888EA 50.77 USD
102539-888
2-Hydroxy-m-anisaldehyde ≥95%
2-Hydroxy-m-anisaldehyde

Hazard: Irritant-Harmful/Stored under Argon/Air Sensitive

Formula: C₈H₈O₃
Boiling Pt: 265…266 °C
Melting Pt: 40…42 °C
MDL Number: MFCD00003322
CAS Number: 148-53-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR