1-Propyl-4-ethynylbenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Ethynyl-4-propylbenzene, (4-Propylphenyl)acetylene, 4-n-Propylphenylacetylene, (4-Propylphenyl)ethyne

075121-1G 075121-5G 075121-10G
102539-864EA 107.69 USD
102539-864 102539-866 102539-868
1-Propyl-4-ethynylbenzene ≥95%
1-Propyl-4-ethynylbenzene

Hazard: Irritant

Formula: C₁₁H₁₂
MDL Number: MFCD00173886
CAS Number: 62452-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR