N-Boc-L-alaninol ≥95%

Supplier: Matrix Scientific

Synonyms: Boc-L-Alaninol, Boc-Alaninol, (S)-2-(BOC-amino)-1-propanol

074955-100G
102539-034EA 258.46 USD
102539-034
N-Boc-L-alaninol ≥95%
N-Boc-L-alaninol

Hazard: Irritant

Formula: C₈H₁₇NO₃
MDL Number: MFCD00043121
CAS Number: 79069-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR