2,4-Dichloro-5-methylpyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2,4-Dichloro-5-picoline

074932-1G 074932-5G
102538-924EA 61.54 USD
102538-924 102538-926
2,4-Dichloro-5-methylpyridine ≥95%
2,4-Dichloro-5-methylpyridine

Hazard: Irritant

Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
MDL Number: MFCD08704813
CAS Number: 56961-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR