2,4-Dichloro-5-methylpyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
2,4-Dichloro-5-picoline
074932-1G
074932-5G
102538-924EA
61.54
USD
102538-924
102538-926
2,4-Dichloro-5-methylpyridine ≥95%
2,4-Dichloro-5-methylpyridine
Hazard: Irritant
Formula:
C₆H₅Cl₂N MW: 162.02 g/mol |
MDL Number:
MFCD08704813 CAS Number: 56961-78-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...