4-Methoxyphenylacetone ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(p-Methoxyphenyl)-2-propanone, 1-(4-Methoxyphenyl)-2-propanone

074922-10G 074922-100G
102538-882EA 64.62 USD
102538-882 102538-884
4-Methoxyphenylacetone ≥95%
4-Methoxyphenylacetone

Hazard: Irritant

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 268
Melting Pt: -15
MDL Number: MFCD00008773
CAS Number: 122-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR