(R)-(-)-Hydratropic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-(-)-2-Phenylpropanoic acid, (R)-(-)-2-Phenylpropionic acid, (R)-2-Phenylpropanoic acid

074907-1G 074907-5G 074907-10G
102538-798EA 143.08 USD
102538-798 102538-800 102538-802
(R)-(-)-Hydratropic acid ≥95%
(R)-(-)-Hydratropic acid

Hazard: Irritant

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 257
MDL Number: MFCD00063140
CAS Number: 7782-26-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR