(R)-(-)-Hydratropic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
(R)-(-)-2-Phenylpropanoic acid, (R)-(-)-2-Phenylpropionic acid, (R)-2-Phenylpropanoic acid
074907-1G
074907-5G
074907-10G
102538-798EA
143.08
USD
102538-798
102538-800
102538-802
(R)-(-)-Hydratropic acid ≥95%
(R)-(-)-Hydratropic acid
Hazard: Irritant
Formula:
C₉H₁₀O₂ MW: 150.18 g/mol Boiling Pt: 257 |
MDL Number:
MFCD00063140 CAS Number: 7782-26-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...