N,O-Bis(trimethylsilyl)acetamide ≥95%

Supplier: Matrix Scientific

Synonyms: BSA, Silyl Derivatisation, Trimethylsilyl N-(trimethylsilyl)acetimidate, Trimethylsilyl N-trimethylsilylacetamidate

074838-100G
102538-480EA 53.85 USD
102538-480
N,O-Bis(trimethylsilyl)acetamide ≥95%
N,O-Bis(trimethylsilyl)acetamide

Hazard: Irritant

Formula: C₈H₂₁NOSi₂
MW: 203.43 g/mol
MDL Number: MFCD00008270
CAS Number: 10416-59-8
UN: 2920
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR