2-(Aminomethyl)benzonitrile hydrochloride ≥95%

Supplier: Matrix Scientific

074778-1G 074778-5G
102538-174EA 384.62 USD
102538-174 102538-176
2-(Aminomethyl)benzonitrile hydrochloride ≥95%
2-(Aminomethyl)benzonitrile hydrochloride

Hazard: Irritant

Formula: C₈H₉ClN₂
MDL Number: MFCD06797973
CAS Number: 1134529-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR