(S)-(+)-1-Aminoindan ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-(+)-1-Indanylamine

074732-5G 074732-10G
102537-930EA 553.85 USD
102537-930 102537-932
(S)-(+)-1-Aminoindan ≥95%
(S)-(+)-1-Aminoindan

Hazard: Irritant

Formula: C₉H₁₁N
MDL Number: MFCD00216670
CAS Number: 61341-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR