(S)-(+)-Camptothecin ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-4-Ethyl-4-hydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, (+)-Camptothecin, 4-Ethyl-4-hydroxy-1H-pyrano-[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione

074699-1G 074699-5G
102537-764EA 76.92 USD
102537-764 102537-766
(S)-(+)-Camptothecin ≥95%
(S)-(+)-Camptothecin

Hazard: Irritant

Formula: C₂₀H₁₆N₂O₄
MW: 348.36 g/mol
Melting Pt: 255…257 °C (dec.)
MDL Number: MFCD00081076
CAS Number: 7689-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR