6-Chloropyridine-2,3-diamine ≥95%

Supplier: Matrix Scientific

074602-1G
102537-272EA 130.77 USD
102537-272
6-Chloropyridine-2,3-diamine ≥95%
6-Chloropyridine-2,3-diamine

Hazard: Toxic

Formula: C₅H₆ClN₃
Melting Pt: 138…140 °C
MDL Number: MFCD00209966
CAS Number: 40851-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR