4-Bromo-2-(trifluoromethoxy)benzaldehyde ≥95%

Supplier: Matrix Scientific

074135-1G 074135-5G 074135-10G
102534-824EA 69.23 USD
102534-824 102534-826 102534-828
4-Bromo-2-(trifluoromethoxy)benzaldehyde ≥95%
4-Bromo-2-(trifluoromethoxy)benzaldehyde

Hazard: Irritant

Formula: C₈H₄BrF₃O₂
MW: 269.0200 g/mol
MDL Number: MFCD12406886
CAS Number: 220996-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR