6-Bromo-4,4-dimethyl-1,4-dihydrobenzo[d][1,3]oxazin-2-one ≥95%

Supplier: Matrix Scientific

074123-1G 074123-5G 074123-10G
102534-758EA 615.38 USD
102534-758 102534-760 102534-762
6-Bromo-4,4-dimethyl-1,4-dihydrobenzo[d][1,3]oxazin-2-one ≥95%
6-Bromo-4,4-dimethyl-1,4-dihydrobenzo[d][1,3]oxazin-2-one

Hazard: Irritant

Formula: C₁₀H₁₀BrNO₂
CAS Number: 21440-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR