3-Benzyloxybenzyl bromide ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(Benzyloxy)benzyl bromide, 1-(Benzyloxy)-3-(bromomethyl)benzene

074074-1G 074074-5G
102534-504EA 196.92 USD
102534-504 102534-506
3-Benzyloxybenzyl bromide ≥95%
3-Benzyloxybenzyl bromide

Hazard: Irritant

Formula: C₁₄H₁₃BrO
MDL Number: MFCD01318307
CAS Number: 1700-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR