(S)-2-(((Benzyloxy)carbonyl)amino)-3-((tert-butoxycarbonyl)amino)propanoic acid ≥95%

Supplier: Matrix Scientific

074072-5G
102534-494EA 200 USD
102534-494
(S)-2-(((Benzyloxy)carbonyl)amino)-3-((tert-butoxycarbonyl)amino)propanoic acid ≥95%
(S)-2-(((Benzyloxy)carbonyl)amino)-3-((tert-butoxycarbonyl)amino)propanoic acid

Hazard: Irritant

Formula: C₁₆H₂₂N₂O₆
MDL Number: MFCD00237342
CAS Number: 16947-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR