(S)-4-Boc-(3-hydroxymethyl)morpholine ≥95%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl (3S)-3-(hydroxymethyl)morpholine-4-carboxylate
073930-250MG
073930-1G
073930-5G
102533-758EA
355.38
USD
102533-758
102533-760
102533-762
(S)-4-Boc-(3-hydroxymethyl)morpholine ≥95%
(S)-4-Boc-(3-hydroxymethyl)morpholine
Hazard: Irritant
Formula:
C₁₀H₁₉NO₄ |
MDL Number:
MFCD06799478 CAS Number: 714971-28-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...