(S)-4-Boc-(3-hydroxymethyl)morpholine ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl (3S)-3-(hydroxymethyl)morpholine-4-carboxylate

073930-250MG 073930-1G 073930-5G
102533-758EA 355.38 USD
102533-758 102533-760 102533-762
(S)-4-Boc-(3-hydroxymethyl)morpholine ≥95%
(S)-4-Boc-(3-hydroxymethyl)morpholine

Hazard: Irritant

Formula: C₁₀H₁₉NO₄
MDL Number: MFCD06799478
CAS Number: 714971-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR