3-Hydroxytetrahydropyran ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Hydroxy-2H-pyran, Tetrahydro-2H-pyran-3-ol

073867-250MG 073867-1G 073867-5G
102533-424EA 2000 USD
102533-424 102533-426 102533-428
3-Hydroxytetrahydropyran ≥95%
3-Hydroxytetrahydropyran

Hazard: Irritant

Formula: C₅H₁₀O₂
MDL Number: MFCD00219387
CAS Number: 19752-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR