4-Iodo-2-methylimidazole ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Iodo-2-methyl-1H-imidazole

073859-1G 073859-5G
102533-380EA 213.85 USD
102533-380 102533-382
4-Iodo-2-methylimidazole ≥95%
4-Iodo-2-methylimidazole

Hazard: Irritant

Formula: C₄H₅IN₂
MW: 208 g/mol
MDL Number: MFCD02684335
CAS Number: 73746-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR