4-Iodo-2-methylimidazole ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Iodo-2-methyl-1H-imidazole
073859-1G
073859-5G
102533-380EA
213.85
USD
102533-380
102533-382
4-Iodo-2-methylimidazole ≥95%
4-Iodo-2-methylimidazole
Hazard: Irritant
Formula:
C₄H₅IN₂ MW: 208 g/mol |
MDL Number:
MFCD02684335 CAS Number: 73746-45-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...