5-Methyl-2-(piperazin-1-yl)thiazole ≥95%

Supplier: Matrix Scientific

073715-250MG 073715-1G 073715-5G
102531-636EA 640 USD
102531-636 102531-638 102531-640
5-Methyl-2-(piperazin-1-yl)thiazole ≥95%
5-Methyl-2-(piperazin-1-yl)thiazole

Hazard: Irritant

Formula: C8H13N3S
CAS Number: 118113-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR