2-Bromoisonicotinonitrile ≥95%
Supplier: Matrix Scientific
073658-5G
073658-25G
102531-328EA
89.23
USD
102531-328
102833-648
2-Bromoisonicotinonitrile ≥95%
2-Bromoisonicotinonitrile
Hazard: Irritant
Formula:
C₆H₃BrN₂ Melting Pt: 70…74 °C |
MDL Number:
MFCD07367879 CAS Number: 10386-27-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...