EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride) ≥95%

Supplier: Matrix Scientific

073580-100G 073580-500G
102530-926EA 67.69 USD
102530-926 102530-928
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride) ≥95%
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride

TCA listed: T
Hazard: Irritant/Moisture Sensitive/Refrigerate/Stored under Argon

Formula: C₈H₁₇N₃·HCl
Melting Pt: ∼113 °C
MDL Number: MFCD00012503
CAS Number: 25952-53-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25d8bf2864bcf21ffe6dcca6f22388d1fe427e22&vid=9080dd6b66a1622ef32a2da2138f85e44cc881b8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR