4-(1-Methylpiperidin-4-yl)aniline ≥95%

Supplier: Matrix Scientific

073481-1G
102530-396EA 415.38 USD
102530-396
4-(1-Methylpiperidin-4-yl)aniline ≥95%
4-(1-Methylpiperidin-4-yl)aniline

Hazard: Irritant

Formula: C₁₂H₁₈N₂
MDL Number: MFCD06804550
CAS Number: 454482-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR