N-Boc-2-amino-5-bromo[1,3,4]thiadiazole ≥95%

Supplier: Matrix Scientific

Synonyms: N-Boc-2-Amino-5-bromo[1,3,4]thiadiazole

073352-1G 073352-5G
102529-714EA 516.92 USD
102529-714 102529-716
N-Boc-2-amino-5-bromo[1,3,4]thiadiazole ≥95%
N-Boc-2-amino-5-bromo[1,3,4]thiadiazole

Hazard: Irritant

Formula: C₇H₁₀BrN₃O₂S
MDL Number: MFCD11848382
CAS Number: 1048358-33-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR